missing translation for 'onlineSavingsMsg'
Learn More

VU0661013, MedChemExpress

Product Code. 16484718
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484718 10 mg 10mg
16471007 50 mg 50mg
16443216 25 mg 25mg
16435546 100 mg 100mg
16427437 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16484718 Supplier Medchem Express Supplier No. HY112859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU661013 is a potent and selective MCL-1 inhibitor.

VU661013 is a potent and selective MCL-1 inhibitor.

Specifications

Chemical Name or Material VU0661013
CAS 2131184-57-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C39H39Cl2N5O4
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (175.40 mM)
SMILES ClC1=CC=C(C(CCCOC2=CC(C)=C(Cl)C(C)=C2)=C3N4[C@H](C)CN(C5=CN(C)C6=C5C=C(C(O)=O)C=C6)C3=O)C4=C1C7=C(C)N(C)N=C7C
Molecular Weight (g/mol) 712.66
Formula Weight 712.66
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.