missing translation for 'onlineSavingsMsg'
Learn More

VUF 11207 fumarate, Tocris Bioscience™

Product Code. 30002060
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30002060

Brand: Tocris Bioscience 4780/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent ACKR3 (CXCR7) agonist

Potent ACKR3 (CXCR7) chemokine receptor agonist (EC50 = 1.6 nM). Induces recruitment of β-arrestin to CXCR7 in HEK293-CXCR7 cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1378524-41-4
Molecular Formula C27H35FN2O4
Molecular Weight (g/mol) 470.59
InChI Key JVRRRKAXHMGUHZ-UHFFFAOYNA-N
Synonym e-n-3-2-fluorophenyl-2-methylallyl-3,4,5-trimethoxy-n-2-1-methylpyrrolidin-2-yl ethyl benzamide, vuf 11207 fumarate, qcr-79, jvrrrkaxhmguhz-xdjhfchbsa-n, n-2e-3-2-fluorophenyl-2-methylprop-2-en-1-yl-3,4,5-trimethoxy-n-2-1-methylpyrrolidin-2-yl ethyl benzamide
PubChem CID 59052285

Specifications

Quantity 10 mg
Formula Weight 586.65
Chemical Name or Material VUF 11207 fumarate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.