missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VUF 11207 fumarate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent ACKR3 (CXCR7) chemokine receptor agonist (EC50 = 1.6 nM). Induces recruitment of β-arrestin to CXCR7 in HEK293-CXCR7 cells.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 586.65 |
| Chemical Name or Material | VUF 11207 fumarate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction