missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 8-Cyclopentyl-1,3-dimethylxanthine
GREENER_CHOICE

Product Code. 30002069
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30002069

Brand: Tocris Bioscience™ 6137/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity adenosine A1 receptor antagonist

8-Cyclopentyl-1,3-dimethylxanthine is a high affinity adenosine A1 receptor antagonist (pKi = 7.5-8).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Cyclopentyl-1,3-dimethylxanthine
CAS 35873-49-5
Synonym 8-Cyclopentyltheophylline
Quantity 100 mg
Solubility Soluble to 100mM in DMSO and to 10mM in ethanol
Molecular Formula C12H16N4O2
Formula Weight 248.28
Physical Form Solid
Color Pale-yellow
Product Type Bioactive Small Molecule
Purity >99%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.