Learn More
Emma Safety Footwear Vulcanus Safety Shoes
Product Code. 15978701
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15978701 43 1 pair
15918701 37 1 pair
15928701 38 1 pair
15938701 39 1 pair
15948701 40 1 pair
15958701 41 1 pair
15968701 42 1 pair
15988701 44 1 pair
15998701 45 1 pair
15908711 46 1 pair
15918711 47 1 pair
15928711 48 1 pair
15938711 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15978701 Supplier Emma Safety Footwear Supplier No. MM53984843
  • ONLINE EXCLUSIVE PRICE
    150.15 €
    93.71 € / 1 pairs
    Save 56.44 €
    37% Off
Estimated Shipment: 24-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Welder's boot designed specifically for working with metal in extreme temperatures. Over-foot flap provides no entry for sparks or shards.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Velcro over-foot flap
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Black
Design Vulcanus
Liner Material Full grain Nappa /Hydro-Tec™ Sanitized Silver
Material Leather
Size 43