Learn More
Emma Safety Footwear Vulcanus Safety Shoes
Description
- S3-Safety shoe with sole plate for wet work
- Full grain leather construction
- GripForce™ Duo Heat Resistant Outsole
- Velcro over-foot flap
- Sanitized
Specifications
Specifications
| Certifications/Compliance | EN ISO 20345:2011 |
| Color | Black |
| Design | Vulcanus |
| Liner Material | Full grain Nappa /Hydro-Tec™ Sanitized Silver |
| Material | Leather |
| Size | 44 |
Product Title
Individuate un'opportunità di miglioramento?