missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Vybrant™ CFDA SE Cell Tracer Kit

Product Code. 10398252
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10398252 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10398252 Supplier Molecular Probes™ Supplier No. V12883
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Vybrant™ CFDA SE Cell Tracer Kit

Vybrant™ CFDA SE Cell Tracer Kit is used in fluorescent analyses as a long-term tracer of cells. This method is commonly used for in vitro and in vivo labeling of cells to determine whether or not a cell is proliferating.

  • Superior performance—allows for bright, uniform staining of cells
  • Long-term retention—dye is well-retained in cells for several days post stain
  • Non-toxic—minimal cytotoxicity for in vivo and in vitro assays
  • Simple, robust staining protocol

    Superior Fluorescent Staining
    Successful tracing of cells requires an extremely bright dye to distinguish fluorescently labeled cells from auto-fluorescence. The intense fluorescent staining provided by Vybrant™ CFDA SE Cell Tracer Kit enables the visualization of labeled cells for several days post stain, making it an ideal stain for cell tracing studies based on dye dilution.

    Long-Term Signal Retention
    Vybrant™ CFDA SE dye easily crosses the plasma membrane and covalently binds inside cells where the stable, well-retained fluorescent dye is designed to provide a consistent signal, even after several days in a cell culture environment.

    Non-Toxic
    Vybrant™ CFDA SE dye binds covalently to all free amines on the surface and inside of cells and shows little cytotoxicity, with minimal observed effect on the proliferative ability or biology of cells. Researchers have used CFDA SE labeling to show that transplantable hematopoietic cells proliferate in vitro in response to stimulation by a growth factor cocktail.

    Simple, Robust Staining Protocol
    The Vybrant™ CFDA SE Cell Tracer Kit contains convenient single-use vials of dry dye to permit small-scale experiments without preparing excess quantities of dye, thereby extending the shelf life of the dye not dissolved in solution. A stock solution is prepared by dissolving the contents of a vial in anhydrous DMSO prior to use. To stain 1 mL of cells in protein-free medium, 1μl of this stock solution is typically used. Cells should be stained for 20 minutes at room temperature with gentle agitation. A brief wash with complete medium will then quench any dye remaining in solution.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Contains 10 vials of 5(6)-CFDA, SE (CFSE, 500 μg per vial), and 1 vial of DMSO (1.3 mL). Store in freezer (-5 to -30°C).
    Excitation Wavelength Range 492⁄517
    Dye Type CFDA SE (CFSE)
    Format Tube(s), Slide(s)
    For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
    Quantity 1 Kit
    Detection Method Fluorescence
    Form Solid
    Product Line Vybrant
    Shipping Condition Room Temperature
    Solubility DMSO (Dimethylsulfoxide)
    Label Type Other Labels or Dyes
    Product Type Tracer
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.