missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Vytac™ Formalin Disposal System, Vytac

Product Code. 10492659 Shop All Epredia Products
Change view
Click to view available options
Description:
Vytac; 1 Gal. (3.79L)
Vytac; 5 Gal. (18.9L)
Unit Size:
Each
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10492659 Vytac; 5 Gal. (18.9L) Each
10240389 Vytac; 1 Gal. (3.79L) Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10492659 Supplier Epredia™ Supplier No. 5405
  • ONLINE EXCLUSIVE PRICE
    196.00 €
    159.19 € / Pack of 1
    Save 36.81 €
    18% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Epredia™ Vytac™ Formalin Disposal System.

Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Includes:
  • Vytac (1 gal.)
  • Reservoir with lid
  • Measuring cup
  • Plastic stir rod
Note: Vytac GNF will neutralize gluteraldehyde
TRUSTED_SUSTAINABILITY

Specifications

Product Type Vytac
Description Vytac; 5 Gal. (18.9L)
Quantity 5 Gal. (18.9 L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.