missing translation for 'onlineSavingsMsg'
Learn More

Memmert™ Wall Bracket for UN55plus

Product Code. 17550316 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
UN55plus Oven
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
17550316 UN55plus Oven Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17550316 Supplier Memmert™ Supplier No. UN55plusB29756

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Wall bracket for wall mounting.

  • Accessory
  • For wall mounting

Specifications

For Use With (Equipment) UN55plus Oven
Type Wall Bracket
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.