Learn More
HALYARD™ Wall Dispenser for boxes of gloves
Product Code. 30274669 Shop All Halyard Products
Change view
Click to view available options
Quantity:
3
Unit Size:
Pack of 3
Product Code. Quantity Unit Size
30274669 3 Pack of 3
1 options
Product Code. 30274669 Supplier HALYARD™ Supplier No. GD001
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Wall Dispensers are a convenient way to hold and organise your boxes of gloves, out of the way but always within reach.
  • Made of clear acrylic
  • Two-way keyholes for wall-mounting
  • Fits most brand name glove boxes
  • Comes in pack of 3 units
Specifications
Product Type Glove Box Holder
Color Clear
Dimensions (L x W x H) 250 x 85 x 132 mm
Holds Disposable Glove Boxes, Exam Glove Boxes, Gloves, Long Cuff Exam Glove Boxes
Material Acrylic
Mounting Wall Mount
No. of Compartments 1
Product Style Side Loading
Capacity 1 Box
Quantity 3
Sterility Non-sterile