Learn More
Ekastu Safety™ Wall Mounting for Eye Wash Bottle
Product Code. 10490043
Change view
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. Material Unit Size
10490043 Stainless Steel Each
1 options
Product Code. 10490043 Supplier Ekastu Safety™ Supplier No. 577950
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides immediate access to an eye wash bottle. EKASTU Safety™ Wall Mounting for Eye Wash Bottle is constructed from high-quality coated steel wire and holds one eye wash bottle.
  • Made of high quality stainless steel
  • Extremely robust and durable
  • Shockproof
  • Easy to install
  • For one eye wash bottle
Specifications
For Use With (Application) Holds One Eyewash Bottle
Material Stainless Steel
Mounting Wall
Product Type Mounting Bracket