missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Buffer A for Dynabeads™ mRNA Purification Kits

Product Code. 15860853
Change view
Click to view available options
Quantity:
1 x 60 mL
Unit Size:
60mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15860853 1 x 60 mL 60mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15860853 Supplier Invitrogen™ Supplier No. A33565
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Wash Buffer A is included in Dynabeads™ mRNA Purification kits.

This Wash Buffer A is included in the following Dynabeads™ mRNA Purification kits (Cat. No. 61011, 61012, and 61021). It is made available separately for applications that require more Wash Buffer A than is provided in the kit.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 60 mL, store (at 2°C to 8°C)
Product Line Dynabeads
Quantity 1 x 60 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.