missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Wash Plate

Product Code. 11982542
Change view
Click to view available options
Quantity:
4/Pk.
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11982542 4/Pk. Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11982542 Supplier Macherey-Nagel™ Supplier No. 740479
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

96‐well plates to facilitate drying of the NucleoSpin 8/96 Binding Strips/Plates and to minimize the risk of cross-contamination.

  • Minimizes the risk of cross-contamination
  • For easy drying
  • Store at 18 to 25°C

Specifications

Description Store at 18 to 25°C
Quantity 4/Pk.
For Use With (Equipment) 96-Well Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.