missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Washing Bottle

Product Code. 10202001 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10202001 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10202001 Supplier Duran™ Supplier No. 247120101
  • ONLINE EXCLUSIVE PRICE
    86.55 €
    75.04 € / Pack of 1
    Save 11.51 €
    13% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Manufactured from borosilicate glass

Manufactured from borosilicate glass

Specifications

Capacity (Metric) 250 mL
Type Gas Washing Bottle
Includes Bottom
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.