missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
221.00 € - 305.00 €
Specifications
| Antigen | WASP |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
WASP Polyclonal antibody specifically detects WASP in Human samples. It is validated for Western BlotSpecifications
| WASP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.0) | |
| 7454 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| IMD2THC, WASP, WiskTHC1, wiskthrombocytopenia 1 (X-linked) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human WASP. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title