Learn More
Kartell SpA Watch Glasses
Product Code. 10356111
Change view
Click to view available options
Diameter (Metric):
60 mm
79 mm
100 mm
120 mm
Unit Size:
Pack of 10
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10356111 100 mm Pack of 10
10061071 60 mm Pack of 10
10746192 79 mm Pack of 10
10448842 120 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10356111 Supplier Kartell SpA Supplier No. 0045500
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Very stable plastic glasses with molded base
  • Polypropylene
  • Autoclavable
Specifications
Autoclavable Autoclavable
Bottom Shape Round
Disposable Reusable
Material Polypropylene
Sterility Non-sterile
Surface Treatment Standard
Type Watch Glass
Diameter (Metric) 100 mm