missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.05% Acetic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Product Code. 15671410
Change view
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
2 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15671410 2 L Glass Bottle 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671410 Supplier Honeywell-Riedel-de Haen Supplier No. 142872L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CHROMASOLV™ LC-MS Ultra, LabReady™ Solvent Blend, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 64-19-7, 7732-18-5
Quantity 2 L
Packaging Glass Bottle
Molecular Formula H2O
UN Number NONH for all modes of transport
Formula Weight 18.02g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Water With 0.05% Acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.