missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.05% Formic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Specifications
Specifications
| CAS | 7732-18-5, 64-18-6 |
| Quantity | 2 L |
| Packaging | Glass Bottle |
| Molecular Formula | H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 18.02g/mol |
| Grade | LC-MS Ultra CHROMASOLV |
| Chemical Name or Material | Water With 0.05% Formic Acid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?