missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.05% Trifluoroacetic Acid (TFA), Optima LC/MS Grade, Fisher Chemical

Product Code. 30117578 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30117578 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30117578 Supplier Fisher Chemical Supplier No. LS1154
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 09-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal high purity mobile phase for LC-MS and UHPLC-UV applications, specifically protein and peptide separations.

  • Suitable for LC-MS and UHPLC-UV
  • Lower TFA content enhances separations by minimizing ion suppression
  • Pre-blended mobile phase simplifies preparation leaving more time to run and analyze samples
  • Tested for the absence of protease that could interfere with protein or peptide analysis
  • TFA is a common mobile phase additive and ion pair reagent in chromatography
TRUSTED_SUSTAINABILITY

Specifications

Grade OPTIMA LC-MS
Quantity 4 L
Form Liquid
Reagent Type Water with 0.05% Trifluoroacetic Acid
CAS 7732-18-5
Packaging Glass Bottle

Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.