missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.1% Formic Acid, LC/MS grade, Thermo Scientific
Shop All Thermo Scientific Products
Click to view available options
Quantity:
2.5 L
4 L
5 L
Unit Size:
2.5 L
4 L
5 L
Description
- Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
- Pre-mixed for ease-of-use
- Low metal ion content to ensure minimal metal contamination
- Formic acid (FA); common mobile phase additive that provides protons for LC/MS analysis
- FA is also known to improve the peak shapes of resulting separation
Specifications
Specifications
| Grade | LC-MS |
| Quantity | 4 L |
| Form | Liquid |
| Reagent Type | Solvent Mixture |
| For Use With | Liquid Chromatography, Mass Spectrometry |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction