missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid, LC/MS grade, Thermo Scientific

Product Code. p-200109924 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2.5 L
4 L
5 L
Unit Size:
2.5 L
4 L
5 L
This item is not returnable. View return policy

Product Code. 30280971

Brand: Thermo Scientific TL01180500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA); common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of resulting separation
TRUSTED_SUSTAINABILITY

Specifications

Grade LC-MS
Quantity 5 L
Form Liquid
Reagent Type Solvent Mixture
For Use With Liquid Chromatography, Mass Spectrometry
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.