missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade

Product Code. 11940379 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
Unit Size:
1 L
2.5 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11940379 4 L Glass Bottle 4 L
10429474 500 mL Glass Bottle 500mL
10229884 1 L Wide Neck Amber Bottle 1 L
10188164 2.5 L Wide Neck Amber Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11940379 Supplier Fisher Chemical Supplier No. LS1184
  • ONLINE EXCLUSIVE PRICE
    202.00 €
    195.68 € / 4 L
    Save 6.32 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA) is a common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of the resulting separation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 3297
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
pH 2
Boiling Point 100°C
Assay Percent Range ≥99.0 %
Merck Index 15, 4269
Grade OPTIMA™ LC-MS
Packaging Glass Bottle
Residue after Evaporation 1ppm max.
Density 1g/cm³
Quantity 4 L
Physical Form Liquid
Chemical Name or Material 0.1% Formic Acid in Water (v/v), Solvent Blends
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.