missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Formic Acid (v/v), Optima™ LC/MS Grade

Product Code. p-3850185 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
Unit Size:
1 L
2.5 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 10429474

Brand: Fisher Chemical LS118500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • Formic acid (FA); common mobile phase additive that provides protons for LC/MS analysis
  • FA is also known to improve the peak shapes of resulting separation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5, 64-18-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 3297
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
pH 2
Boiling Point 100°C
Assay Percent Range ≥99.0 %
Merck Index 15, 4269
Grade OPTIMA™ LC-MS
Packaging Glass Bottle
Residue after Evaporation 1ppm max.
Density 1g/cm³
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material 0.1% Formic Acid in Water (v/v), Solvent Blends
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.