missing translation for 'onlineSavingsMsg'
Learn More

Water with 0.1% Trifluoroacetic Acid (v/v), Optima™ LC/MS Grade, Thermo Scientific™

Product Code. 10311725 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Wide Neck Amber Bottle
Unit Size:
1 L
2.5 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10311725 1 L Wide Neck Amber Bottle 1 L
10362055 500 mL Glass Bottle 500mL
10516625 2.5 L Wide Neck Amber Bottle 2.5 L
11900379 4 L Glass Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10311725 Supplier Fisher Chemical Supplier No. LS1191
  • ONLINE EXCLUSIVE PRICE
    89.15 €
    58.60 € / 1 L
    Save 30.55 €
    34% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Ideal high purity mobile phase blend for LC/MS and UHPLC-UV applications
  • Pre-mixed for ease-of-use
  • Low metal ion content to ensure minimal metal contamination
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5 , 64-18-6
PubChem CID 962
ChEBI CHEBI:15377

Specifications

Absorbance Report AU at 210nm, Report AU at 220nm, Report AU at 230nm, Report AU at 254nm
Assay Percent Range 99.0 to 103.0 %
Molecular Formula H2O
MDL Number 4169
Merck Index 15, 9847
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Percent Purity 0.095 to 0.105% (v/v) Trifluoroacetic Acid
Grade OPTIMA™ LC-MS
Packaging Wide Neck Amber Bottle
Residue after Evaporation ≤1ppm
Quantity 1 L
Physical Form Liquid
Chemical Name or Material 0.1% Trifluoroacetic Acid in Water (v/v), Solvent Blends
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.