missing translation for 'onlineSavingsMsg'
Learn More

Water, ACS reagent

Product Code. 10433160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Plastic Bottle
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10433160 1 L Plastic Bottle 1 L
10892431 5 L Plastic Bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10433160 Supplier Thermo Scientific Acros Supplier No. 434400010
  • ONLINE EXCLUSIVE PRICE
    50.70 €
    47.00 € / 1 L
    Save 3.70 €
    7% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Absorbance Passes Test, 0.01 max. at 210nm, 0.005 max. at 254nm, 0.01 max. at 200nm, 0.005 max. at 300nm
Color White
Boiling Point 100°C
Conductivity 2μS/cm Max.
Linear Formula H2O
Merck Index 15,10237
Formula Weight 18.02
Grade ACS Reagent
Packaging Plastic Bottle
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Crystalline Powder
Chemical Name or Material Water, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.