missing translation for 'onlineSavingsMsg'
Learn More

Water, for analysis

Product Code. 10543245 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10543245 2.5 L Plastic Bottle 2.5 L
10530194 1 L Plastic Bottle 1 L
11914391 10 L Plastic Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10543245 Supplier Thermo Scientific Acros Supplier No. 389400025
  • ONLINE EXCLUSIVE PRICE
    47.30 €
    43.90 € / 2.5 L
    Save 3.40 €
    7% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.01 max. (water, 1cm, 254nm)
pH 5 to 8
Color Colorless
Boiling Point 100.0°C
Conductivity 6μS/cm Max.
Linear Formula H2O
Formula Weight 18.02
Grade Analysis
Packaging Plastic Bottle
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 2.5 L
Refractive Index 1.3334
Specific Gravity 1
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.