missing translation for 'onlineSavingsMsg'
Learn More

Water, BAKER ANALYZED™, J.T. Baker™

Product Code. 15518664 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15518664 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518664 Supplier JT Baker Supplier No. 9825.1000GL
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
    • at 210 nm max. 0.005
    • at 254 nm max. 0.001
  • LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
    • Positive ESI-MS Sensitive Impurities (as Reserpine) max. 100 ng/mL

Specifications

CAS 7732-18-5
Melting Point 0°C
Boiling Point 100°C (1013 hPa)
Molecular Formula H2O
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Packaging Glass Bottle
Residue after Evaporation max. 5 ppm
Density 1 g/cm³ (4°C)
Quantity 1 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.