missing translation for 'onlineSavingsMsg'
Learn More

Water, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 10546602 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10546602 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10546602 Supplier JT Baker Supplier No. 4218.2500
  • ONLINE EXCLUSIVE PRICE
    64.00 €
    54.20 € / 2.5 L
    Save 9.80 €
    15% Off
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HPLC gradient grade water for use in Liquid Chromatography (HPLC & UHPLC) and Spectrophotometry, packaged in 2.5L glass bottles.

BAKER ANALYZED™ HPLC grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Manufactured and tested to assure interference-free separations
  • Highly characterized reagents that have unmatched lot-to-lot consistency
  • Minimizes recalibration or instrument adjustments
  • Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
  • An Actual Lot Analysis is provided on the label

Specifications

Absorbance Max. 0.005 au at 220nm, Max. 0.001 au at 254nm
pH 7
CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
Grade BAKER ANALYZED™ HPLC
Odor Odorless
Packaging Glass Bottle
Residue after Evaporation Max. 2ppm
Vapor Pressure 0.05 kPa
Density 1.00g/mL
Quantity 2.5 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.