missing translation for 'onlineSavingsMsg'
Learn More

Water, BAKER INSTRA-ANALYZED Plus, J.T. Baker™

Product Code. 15578594 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578594 1 L Glass 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578594 Supplier JT Baker Supplier No. 9381.1000
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Analyzed for up to 64 metals in the low parts-per-billion (ppb) range, ensuring low background interference for conducting critical trace metal analysis.

BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.

  • Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
  • Lot to lot consistency ensures reliable results between tests
  • Space-saving HDPE packaging with low metallic extractables
  • Enhanced safety with tamper-evident caps
  • Environmentally-friendly recyclable HDPE bottles
  • Strict protocols in an ISO facility ensure quality in manufacturing

Specifications

Absorbance Max. 0.01 at 200nm, Max. 0.005 at 254nm, Max. 0.005 at 300nm, Max. 0.005 at 400nm
CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02g/mol
Grade For use in Trace Metal Analysis
Odor Odorless
Packaging Glass
Residue after Evaporation Max. 1 ppm
Viscosity 0.952 mPas
Density 1g/cm3
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.