missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Water, Cell Culture Grade

Product Code. 31908413 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31908413 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31908413 Supplier Sartorius Supplier No. 030551A
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

It undergoes pretreatment and is then produced by Reverse Osmosis (RO) and Deionization (DI) processes.

  • This cell culture grade water is continually monitored for endotoxins, microbial bioburden and metals, in order to ensure that all international acceptable specifications are met.

Specifications

Chemical Name or Material Water
Molecular Weight (g/mol) Unknown
Percent Purity Unknown
Grade Cell Culture
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.