missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva

Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

Brand:  HyClone™ SH30529.03

Code : 83

Additional Details : Weight : 1.28160kg

Quantity 1L

 View more versions of this product

Product Code. 10011342

  • 15.72 € / 1000mL


In stock, ready to ship: 89
Add to basket

Description

Description

  • Endotoxin-free; contains no detectable gram-negative bacteria
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
Specifications

Chemical Identifiers

5.0 to 7.0

Specifications

0.1μm sterile filtered
1L
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
5.0 to 7.0
Videos
SDS
Documents