missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Water, Cell Culture Grade (Endotoxin-Free), Cytiva

Product Code. 10185873
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1000mL
100mL
500mL
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10185873 6 x 1 L Pack of 6
10183152 100 mL 100mL
10001342 500 mL 500mL
10011342 1 L 1000mL
11586271 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10185873 Supplier HyClone products Cytiva Supplier No. SH30529.LS
  • ONLINE EXCLUSIVE PRICE
    175.78 €
    135.00 € / Pack of 6
    Save 40.78 €
    23% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eliminate endotoxin concerns with HyClone™ Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.

  • Endotoxin-free; contains no detectable gram-negative bacteria
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
TRUSTED_SUSTAINABILITY

Specifications

Quantity 6 x 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.