missing translation for 'onlineSavingsMsg'
Learn More

Water, Demineralized, Chem-Lab

Product Code. 11918514
Change view
Click to view available options
Quantity:
10 L
25 L
Unit Size:
10 L
25 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11918514 25 L 25 L
11969242 10 L 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11918514 Supplier Chem-Lab Supplier No. CL02.2102.9025
  • ONLINE EXCLUSIVE PRICE
    86.00 €
    41.40 € / 25 L
    Save 44.60 €
    51% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for laboratory use. Water, Demineralized, Chem-Lab is purified to remove minerals and salts with LF <5μS. There are less than 5ppm of non-volatile matter. Available in 10 and 25L cartons.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH ±7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.016g/mol
Odor Odorless
Filtered Through LF <5μS
Density 1.00g/mL
Quantity 25 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.