Learn More
Heidolph™ Water Distribution Manifold
Product Code. 11741709 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
Starfish Magnetic Stirrer System.
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
11741709 Starfish Magnetic Stirrer System. Each
1 options
Product Code. 11741709 Supplier Heidolph™ Supplier No. 5058103000
  • ONLINE EXCLUSIVE PRICE
    916.00 €
    906.00 € / Pack of 1
    Save 10.00 €
    1% Off
Estimated Shipment: 01-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Distribute coolant from a single source evenly. Heidolph™ Water Distribution Manifold can evenly distribute to up to five condensers and then recombine the flow to one outlet pipe.
  • Distribute to up to 5 condensers.
  • Recombines flow to one outlet pipe.
  • Leak-proof shut-off valves.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Starfish Magnetic Stirrer System.
For Use With (Application) Water Distribution.
Includes Leak-proof shut-off valves.