missing translation for 'onlineSavingsMsg'
Learn More

Water, (DNASE, RNASE free), Fisher BioReagents™

Product Code. 10336503
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Nalgene Poly Bottle
Poly Bottle
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336503 100 mL Poly Bottle 100mL
10295243 50 mL Nalgene Poly Bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336503 Supplier Fisher BioReagents Supplier No. BP2484100
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water, H2O, CAS Number-7732-18-5, dihydrogen monoxide, dihydrogen oxide, 100mL, Vapor Pressure: 17.5mmHg at 20 deg.C, 100 deg.C, CHEBI:15377, Gray, Colorless, 1.000g/cm3, DNase free, Filtered through a 0.2-micron filter., 18.02, Low hazard for usual industrial or commercial handling.

Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Grade Molecular Biology
Packaging Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Treatment(s) DEPC-Treated
Density 1.000g/cm³
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Water, DNASE, RNASE free
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.