missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Water, Endotoxin-free

Product Code. p-7043114
Change view
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456979 500 mL 500mL
15466979 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456979 Supplier Thermo Scientific Chemicals Supplier No. J65589.AP
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Water Endotoxin-free is used as solvent in industries and in all research fields. It is also used in laboratory for cell isolation and culture. It is also useful for elution of endotoxin- free plasmid DNA.

Solubility
Miscible with methanol, acetone and ethanol.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Melting Point 32°F
Boiling Point 100°C
Beilstein 2050024
Merck Index 1410039
Solubility Information Miscible with methanol,acetone and ethanol.
Formula Weight 18.02
Grade Endotoxin-Free
Odor Odorless
Density 1
Quantity 500 mL
Refractive Index 1.34
Physical Form Liquid
Chemical Name or Material Water, Endotoxin-free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.