missing translation for 'onlineSavingsMsg'
Learn More

Water For Ion Chromatography, Fisher Chemical™
BIOPHARMA

Product Code. 12993634 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
2000 mL
Unit Size:
1000mL
2000mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12993634 1000 mL 1000mL
12903644 2000 mL 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12993634 Supplier Fisher Chemical Supplier No. W 001 1000
  • ONLINE EXCLUSIVE PRICE
    96.40 €
    70.24 € / 1000mL
    Save 26.16 €
    27% Off
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 1000 mL
Chemical Name or Material Water for Ion Chromatography
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.