missing translation for 'onlineSavingsMsg'
Learn More

Water, GC Headspace Grade, Thermo Scientific™

Product Code. 15552233 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
Packaging:
Glass Bottle
missing translation for 'unitSize':
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
15552233 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15552233 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' W101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water processed to high purity for accurate and repeatable determination of trace levels of residual solvents

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
TRUSTED_SUSTAINABILITY

Specifications

Absorbance 0.005 AU Maximum (400 nm), 0.005 AU Maximum (300 nm), 0.005 AU Maximum (280 nm), 0.005 AU Maximum (250 nm), 0.010 AU Maximum (220 nm), 0.010 AU Maximum (190 nm)
CAS 7732-18-5
Molecular Formula H2O
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.01
Grade GC Headspace
Packaging Glass Bottle
Residue after Evaporation 1 ppm max
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.