Learn More
WTW™ Water Hardness Test Kits
Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits
Brand: WTW™ 250404
Code : A0
Additional Details : Weight : 0.80000kg
Specifications
Phthalein complexone | |
PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral photometers | |
25 | |
25 Tests | |
RH residual hardness cell test kit |
To check of ion-exchanger plants for the production of deionized water | |
Certificates | |
0.075 to 0.750 °d, 0.5 to 5 mg/L Ca | |
Photometric |
Safety and Handling
- residual hardness cell test method
Signal Word
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide