missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Water Hardness Test Kits

Product Code. p-8002898
Click to view available options
Type:
RH Residual Hardness
Water Hardness, Total
Unit Size:
25g
Pack of 25
This item is not returnable. View return policy

Product Code. 11732285

Brand: WTW™ 252039

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Test materials for the AQA of photometric routine and water analysis for multi-level result certainty of photoLab™, pHotoFlex™ and test kits

TRUSTED_SUSTAINABILITY

Specifications

Analytes Phthalein complexone
For Use With (Application) For measurement of Water hardness, total hardness
For Use With (Equipment) PhotoLab™ S6, PhotoLab™ S12, PhotoLab™ Spectral, Photoflex photometers
Includes Certificates
No. of Tests 25
Range 0.7 to 30.1 °d, 5 to 215 mg/L Ca
Sufficient For 25 Tests
Test Method Photometric
Type Water Hardness, Total
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.