missing translation for 'onlineSavingsMsg'
Learn More

Water, Solstice

Produktkod. 17854591
Change view
Klicka för att se tillgängliga alternativ
Quantity:
1 L
6 x 1 L
Packaging:
Glass Bottle
Förpackningsstorlek:
1 L
6 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Packaging Förpackningsstorlek
17854591 6 x 1 L Glass Bottle 6 L
15665760 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17854591 Leverantör Honeywell Fluka Leverantörsnummer 534636X1L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CHROMASOLV™, GC-Headspace tested

Solstice Chromasolv™ GC-headspace solvents:

  • developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
  • very pure solvents with extremely low concentrations of the defined residual solvents
  • state-of-the art production and application tests
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 7732-18-5
Molecular Formula H2O
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifikationer

UN Number NONH for all modes of transport
Beilstein 2050024
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor
IUPAC Name water
Molecular Weight (g/mol) 18.02
Formula Weight 18.02g/mol
Packaging Glass Bottle
Density 1.000g/mL at 3.98°C
Quantity 6 x 1 L
Chemical Name or Material Water
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.