missing translation for 'onlineSavingsMsg'
Learn More

Water, for HPLC gradient grade

Product Code. 10596261 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10596261 1 L 1 L
10043370 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10596261 Supplier Thermo Scientific Supplier No. 268300010
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5, H2O, 18.015 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100.0°C
Linear Formula H2O
Fieser 16,383
Merck Index 15, 10237
Formula Weight 18.02
Grade HPLC Gradient
Packaging Glass Bottle
Residue after Evaporation 0.0003% max.
Vapor Pressure 17.5mmHg at 20°C
Viscosity 0.89 mPa.s (25°C)
Density 1.0000g/mL
Quantity 1 L
Refractive Index 1.3334
Specific Gravity 1
Green Features ACT Ecolabel (Gold)
Chemical Name or Material Water, For HPLC gradient grade
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.