missing translation for 'onlineSavingsMsg'
Learn More

Water for Injection, USP, Sterile Grade, Cytiva Hyclone™

Product Code. 31937776
Change view
Click to view available options
Quantity:
6 x 1 L
Packaging:
PET Plastic Bottle
Unit Size:
6 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31937776 6 x 1 L PET Plastic Bottle 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31937776 Supplier HyClone products Cytiva Supplier No. WFIUSP1X6
  • ONLINE EXCLUSIVE PRICE
    73.05 €
    90.59 € / 6 L
    Save POA
    -24.01% Off
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Water for Injection, USP, is a terminally distilled, sterile, non-pyrogenic preparation which contains no bacteriostat, antimicrobial agent or added buffer.

  • Complies with the USP monograph for water for injection packaged in bulk for commercial use
  • Manufactured under cGMP and ISO 13485 guidelines in a FDA registered facility
  • Excellent choice for upstream and downstream media/buffer preparation in biopharmaceutical manufacturing applications
  • Widely used in applications where a high quality sterile water is required

For Manufacturing or Research Use only, not intended for Diagnostic or Therapeutic Use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Clear, Colorless
Endotoxin <0.25 EU/mL
Grade USP
Packaging PET Plastic Bottle
Sterility Sterile
Certificate of Analysis Yes
Quantity 6 x 1 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.