missing translation for 'onlineSavingsMsg'
Learn More

Water, CHROMASOLV™ LC-MS, Solstice

Product Code. 15608400
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
4 L
4 x 4 L
Unit Size:
1 L
2.50 L
250mL
4 L
500mL
Each
Pack of 4
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15608400 4 L 4 L
15685350 250 mL 250mL
15605360 500 mL 500mL
15665350 1 L 1 L
16225301 2.5 L 2.50 L
16419019 2.5 L Each
15695350 4 x 4 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 15608400 Supplier Honeywell-Chromasolv Supplier No. 392534L
  • ONLINE EXCLUSIVE PRICE
    128.00 €
    81.88 € / 4 L
    Save 46.12 €
    36% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CHROMASOLV™ LC-MS

  • For LC-MS and LC-UV trace metals analysis
  • HPLC / UV-VIS quality control
  • Low gradient baseline drift
  • Low levels of trace metals
  • No non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Absorbance max. 0.022 at 210 nm, max. 0.004 at 230 nm
Boiling Point 100°C
CAS Max % 100.0000%
Linear Formula H2O
Beilstein 2050024
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Formula Weight 18
Grade LC-MS
Packaging Glass Bottle
Density 1.000 g/mL at 3.98 °C(lit.)
Quantity 4 L
Chemical Name or Material Water, CHROMASOLV™ LC-MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.