missing translation for 'onlineSavingsMsg'
Learn More

Water, LC/MS grade, Thermo Scientific

Product Code. 30280924 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280924 2.5 L 2.5 L
30280922 500 mL 500mL
30280923 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30280924 Supplier Thermo Scientific Supplier No. T000062500
  • ONLINE EXCLUSIVE PRICE
    135.00 €
    92.90 € / 2.5 L
    Save 42.10 €
    31% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Weight (g/mol) 18.02 g/mol

Specifications

CAS 7732-18-5
Percent Purity 99.90%
Grade LC-MS
Packaging Glass Bottle
Quantity 2.5 L
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.