missing translation for 'onlineSavingsMsg'
Learn More

Water, Microbial Cell Culture Grade, Fisher BioReagents

Product Code. 11373892
Change view
Click to view available options
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11373892 1 L Nalgene Poly Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11373892 Supplier Fisher BioReagents Supplier No. BP28201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Ultrapure water for preparing microbiological growth media.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Conductivity 2μS/cm Max.
Endotoxin 0.025 EU/mL max.
Formula Weight 18.01
Grade Microbial Cell Culture
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through 0.03 Micron filtered and autoclaved
Density 1.000g/cm³
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water, 0.03μm filtered and autoclaved
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.