missing translation for 'onlineSavingsMsg'
Learn More

Melford Water Molecular Biology Grade DNase and RNase Free

Product Code. 30968823
Change view
Click to view available options
Quantity:
1 L
1 gal.
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30968823 1 gal. 4 L
30017924 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30968823 Supplier Melford Supplier No. 2487004000.0
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified RNase and DNase free for use in molecular biology applications.

Certified RNase and DNase free for use in molecular biology applications. Ideal for making reagents, rinsing glass and plastic ware, enzymatic reactions and more. Toxic agents such as DEPC are not used in purification procedures.

  • Endotoxin Free
  • RNase Free
  • DNase Free

Tested to < 0.001 EU/ML

Meets or exceeds CAP/CLSI specifications for Clinical Laboratory Reagent Water (CLRW) and USP/NF Purified Water.

Specifications

Chemical Name or Material Water, Molecular Biology Grade, DNase and RNase Free
CAS 7732-18-5
Molecular Weight (g/mol) 18
Grade Molecular Biology
Quantity 1 gal.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.