missing translation for 'onlineSavingsMsg'
Learn More

Water, Molecular Biology Grade, Fisher BioReagents

Product Code. 10517554
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
10 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
Unit Size:
1 L
10 L
100mL
20 L
4 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10517554 20 L Poly CUBE 20 L
10490025 100 mL Poly Bottle 100mL
10505854 1 L Nalgene Poly Bottle 1 L
10154604 4 L Poly CUBE 4 L
10318354 10 L Poly CUBE 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10517554 Supplier Fisher BioReagents Supplier No. BP281920
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For many fundamental molecular biology procedures

Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity <2μS/cm
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Grade DNase-, RNase- and Protease-Free
Packaging Poly CUBE
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.03μ filter
Treatment(s) Deionized
Density 1.000g/cm³
Quantity 20 L
Physical Form Liquid
Chemical Name or Material Water, 0.03μm filtered to ensure high purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.