missing translation for 'onlineSavingsMsg'
Learn More

Water, Nuclease-free, Molecular Biology Grade, Ultrapure

Product Code. 15845408 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
10 x 1 mL
500 mL
1 L
5 L
Packaging:
Nalgene Bottle
Upright carton
Unit Size:
1 L
100mL
10mL
5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15845408 5 L Upright carton 5 L
15644028 10 x 1 mL Nalgene Bottle 10mL
15815408 100 mL Nalgene Bottle 100mL
15825408 500 mL Nalgene Bottle 500mL
15835408 1 L Nalgene Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15845408 Supplier Thermo Scientific Supplier No. J71786.K8
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7732-18-5 | H2O | 18.015 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Molecular Biology
Packaging Upright carton
Density 1
Quantity 5 L
Specific Gravity 1.000
Physical Form Liquid
Chemical Name or Material Water

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.