missing translation for 'onlineSavingsMsg'
Learn More

Elgas™ Water Purifier

Product Code. 13105970
Change view
Click to view available options
Flow Rate:
15 L/hr.
7 L/hr.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate unitSize
13105970 15 L/hr. Each
13195960 7 L/hr. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13105970 Supplier Elgas™ Supplier No. PRIPLB0236
  • ONLINE EXCLUSIVE PRICE
    10783.50 €
    8731.62 € / Pack of 1
    Save 2051.88 €
    19% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Elgas™ Water Purifier uses Purelab™ Option-S Series multiple purification technologies to produce general laboratory grade water at better than single distilled quality.

  • Purification technologies include pre-treatment, reverse osmosis, and ion exchange.
  • Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets.

Specifications

Conductivity 1400μS/cm
Flow Rate 15 L/hr.
Temperature Range (Metric) 1°C to 35°C
Width (Metric) 410 mm
Height (Metric) 460 mm
Depth (Metric) 270 mm
Weight (Metric) 14.5 kg
Electrical Requirements 100/240 V 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Product Type Water Purifier
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.